Dantrolene sodium salt hydrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 1-[5-(4-Nitrophenyl)furfurylideneamino]hydantoin Sodium Salt

D3996-1G D3996-5G
TCIAD3996-1GEA 45 EUR
TCIAD3996-1G TCIAD3996-5G
Dantrolene sodium salt hydrate ≥98.0% (by HPLC, titration analysis)
Dantrolene sodium salt hydrate
Formula: C₁₄H₁₀N₄O₅
MW: 336,24 g/mol
Número MDL: MFCD00079130
Núm. CAS: 14663-23-1
EINECS: 238-706-8
Índice Merck: 14,02816
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=151b60e5835e7a418998c83ea6d37afe0f789089&vid=94a5c2a04c210522f142cf23b367c79ba300e4d8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 20.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR