(S)-(-)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(S)-2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene, (S)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine, (S)-3-(Dimethylamino)-1-(2-thienyl)-1-propanol
D4010-1G
D4010-5G
TCIAD4010-1GEA
16
EUR
TCIAD4010-1G
TCIAD4010-5G
(S)-(-)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine ≥98.0% (by GC, titration analysis)
(S)-(-)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine
Formula:
C₉H₁₅NOS MW: 185.2865 g/mol |
Número MDL:
MFCD07782107 Núm. CAS: 132335-44-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Specific rotation [a]20/D | -4.0 to -8.0 deg (C=1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...