Di(N-succinimidyl) sebacate ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: Sebacic acid bis(N-succinimidyl) ester, Di(N-succinimidyl) Decanedioate, Sebacic acid Di(N-succinimidyl) ester, Decanedioic acid Di(N-succinimidyl) ester, Bis(2,5-dioxopyrrolidin-1-yl) decanedioate

D4019-1G D4019-5G
TCIAD4019-1GEA 51 EUR
TCIAD4019-1G TCIAD4019-5G
Di(N-succinimidyl) sebacate ≥98.0% (by HPLC, total nitrogen)
Di(N-succinimidyl) sebacate
Formula: C₁₈H₂₄N₂O₈
MW: 396.40 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00216005
Núm. CAS: 23024-29-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR