2,6-Dichlorocinnamic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-(2,6-Dichlorophenyl)acrylic acid, (2E)-3-(2,6-Dichlorophenyl)acrylic acid
D4048-5G
D4048-25G
TCIAD4048-5GEA
24
EUR
TCIAD4048-5G
TCIAD4048-25G
2,6-Dichlorocinnamic acid ≥98.0% (by GC, titration analysis)
2,6-Dichlorocinnamic acid
Formula:
C₉H₆Cl₂O₂ MW: 217,05 g/mol |
Número MDL:
MFCD00004374 Núm. CAS: 5345-89-1 EINECS: 226-301-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...