Docetaxel ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
D4102-100MG
D4102-1G
TCIAD4102-100MGEA
59
EUR
TCIAD4102-100MG
TCIAD4102-1G
Docetaxel ≥98.0% (by HPLC)
Docetaxel
Formula:
C₄₃H₅₃NO₁₄ |
Número MDL:
MFCD00800737 Núm. CAS: 114977-28-5 Índice Merck: 14,03397 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -42.0 to -45.0 deg (C=1、EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...