Diflufenican ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: N-(2,4-Difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]nicotinamide

D4108-5G D4108-25G D4108-1G
TCIAD4108-5GEA 85 EUR
TCIAD4108-5G TCIAD4108-25G TCIAD4108-1G
Diflufenican ≥98.0% (by HPLC, total nitrogen)
Diflufenican
Formula: C₁₉H₁₁F₅N₂O₂
MW: 394,30 g/mol
Número MDL: MFCD01311804
Núm. CAS: 83164-33-4
Índice Merck: 14,03144

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 159.0 to 163.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR