Disodium clodronate tetrahydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Clodronic Acid Disodium Salt, Clodronate disodium tetrahydrate, Sodium (dichloro(phosphono)methyl)phosphonate tetrahydrate

D4160-1G D4160-5G
TCIAD4160-1GEA 65 EUR
TCIAD4160-1G TCIAD4160-5G
Disodium clodronate tetrahydrate ≥98.0% (by titrimetric analysis)
Disodium clodronate tetrahydrate
Formula: CH₄Cl₂O₆P₂
MW: 288,85 g/mol
Número MDL: MFCD02940846
Núm. CAS: 88416-50-6
Índice Merck: 14,02371

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 18.0 to 22.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR