Dimethylphenylphosphine borane ≥95.0% (by HPLC)

Proveedor: TCI

Sinónimos: Borane - Dimethylphenylphosphine Complex

D4204-1G D4204-5G
TCIAD4204-1GEA 86 EUR
TCIAD4204-1G TCIAD4204-5G
Dimethylphenylphosphine borane ≥95.0% (by HPLC)
Dimethylphenylphosphine borane
Formula: C₈H₁₄BP
MW: 151,98 g/mol
Pto. Ebullición: 78°C
Núm. CAS: 35512-87-9

Order Now

Resultados test especificaciones

Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR