Di-tert-butyl hydrazodicarboxylate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: Di-tert-butyl hydrazine-1,2-dicarboxylate, Di-tert-butyl Hydrazodiformate, Di-tert-butyl 1,2-Hydrazinedicarboxylate

D4224-5G D4224-25G
TCIAD4224-5GEA 23 EUR
TCIAD4224-5G TCIAD4224-25G
Di-tert-butyl hydrazodicarboxylate ≥98.0% (por GC)
Di-tert-butyl hydrazodicarboxylate
Formula: C₁₀H₂₀N₂O₄
MW: 232,28 g/mol
Número MDL: MFCD00015000
Núm. CAS: 16466-61-8
EINECS: 240-512-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR