2,5-Dimethylphenylacetyl chloride ≥98.0% (by GC, titration analysis)
Proveedor: TCI
D4261-5G
D4261-25G
TCIAD4261-5GEA
29
EUR
TCIAD4261-5G
TCIAD4261-25G
2,5-Dimethylphenylacetyl chloride ≥98.0% (by GC, titration analysis)
2,5-Dimethylphenylacetyl chloride
Resultados test especificaciones
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...