2,3-Dihydrobenzofuran-5-acetic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-Coumaranacetic Acid, 2,3-Dihydrobenzofuranyl-5-acetic acid

D4270-1G D4270-5G
TCIAD4270-1GEA 29 EUR
TCIAD4270-1G TCIAD4270-5G
2,3-Dihydrobenzofuran-5-acetic acid ≥98.0% (by GC, titration analysis)
2,3-Dihydrobenzofuran-5-acetic acid
Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Número MDL: MFCD01941301
Núm. CAS: 69999-16-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR