5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
5,6-Dichloro-1-(β-D-ribofuranosyl)benzimidazole, 5,6-Dichlorobenzimidazole Riboside, (2R,3R,4S,5R)-2-(5,6-Dichloro-1H-benzo[d]imidazol-1-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, DRB
D4292-100MG
D4292-1G
TCIAD4292-100MGEA
72
EUR
TCIAD4292-100MG
TCIAD4292-1G
5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside ≥98.0% (by HPLC, total nitrogen)
5,6-Dichlorobenzimidazole-1-beta-D-Ribofuranoside
Formula:
C₁₂H₁₂Cl₂N₂O₄ MW: 319.14 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00036785 Núm. CAS: 53-85-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 220.0 to 224.0 deg-C |
Specific rotation [a]20/D | -66.0 ~ -68.0 deg (C=2, Pyridine) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...