Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
[4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]palladium(II) Dichloride, [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]dichloropalladium(II)
D4333-200MG
D4333-1G
TCIAD4333-200MGEA
28
EUR
TCIAD4333-200MG
TCIAD4333-1G
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) ≥98.0% (by titrimetric analysis)
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)
Formula:
C₃₉H₃₂Cl₂OP₂Pd MW: 755,95 g/mol |
Número MDL:
MFCD14155707 Núm. CAS: 205319-10-4 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...