(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(S)-N-Despropyl Pramipexole, Pramipexole Related Compound A
D4337-1G
D4337-5G
TCIAD4337-1GEA
51
EUR
TCIAD4337-1G
TCIAD4337-5G
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole ≥98.0% (by GC, titration analysis)
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole
Formula:
C₇H₁₁N₃S MW: 169,25 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD07368003 Núm. CAS: 106092-09-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 227.0 to 231.0 deg-C |
Specific rotation [a]20/D | -94.0 to -98.0 deg (C=1,MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...