N,N-Dimethyl-N-(methylsulphanylmethylene)ammonium iodide ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N,N-Dimethyl-N-(methylsulphanylmethylene)iminium Iodide
D4340-5G
D4340-25G
TCIAD4340-5GEA
64
EUR
TCIAD4340-5G
TCIAD4340-25G
N,N-Dimethyl-N-(methylsulphanylmethylene)ammonium iodide ≥98.0% (by titrimetric analysis)
N,N-Dimethyl-N-(methylsulphanylmethylene)ammonium iodide
Formula:
C₄H₁₀INS MW: 231,10 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD17015287 Núm. CAS: 29085-13-0 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...