Dibenzothiophene-2-boronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
Dibenzo[b,d]thiophen-2-ylboronic acid
D4373-1G
D4373-5G
TCIAD4373-1GEA
45
EUR
TCIAD4373-1G
TCIAD4373-5G
Dibenzothiophene-2-boronic acid (contains varying amounts of Anhydride)
Dibenzothiophene-2-boronic acid
Formula:
C₁₂H₉BO₂S MW: 228,07 g/mol |
Número MDL:
MFCD01318982 Núm. CAS: 668983-97-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 95.0 to 109.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...