Chalcone-α,β-epoxide ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
Chalcone Oxide, 2-Benzoyl-3-phenyloxirane, 1,3-Diphenyl-2,3-epoxy-1-propanone, 1,3-Diphenyl-2-propen-1-one Oxide
D4378-1G
D4378-5G
TCIAD4378-1GEA
16
EUR
TCIAD4378-1G
TCIAD4378-5G
Chalcone-α,β-epoxide ≥98.0% (por GC)
Chalcone-alpha,beta-epoxide
Formula:
C₁₅H₁₂O₂ MW: 224.26 g/mol |
Número MDL:
MFCD00022336 Núm. CAS: 5411-12-1 EINECS: 226-487-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...