3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

D4456-200MG D4456-1G
TCIAD4456-200MGEA 63 EUR
TCIAD4456-200MG TCIAD4456-1G
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥98.0% (by total nitrogen basis)
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]
Formula: C₂₅H₂₇IN₂S₂
Número MDL: MFCD00075033
Núm. CAS: 53213-94-8

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR