3,3,3',3'-Tetramethyl-1,1'-bis(4-sulphobutyl)benzoindodicarbocyanine sodium salt ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: Sodium 4-[2-[5-[1,1-Dimethyl-3-(4-sulphonatobutyl)-1H-benz[e]indol-2(3H)-ylidene]-1,3-pentadienyl]-1,1-dimethyl-1H-benz[e]indol-3-ium-3-yl]butane-1-sulphonate, NK 1841, 5-[1,1-Dimethyl-3-(4-sulphonatobutyl)benz[e]indolin-2-ylidene]-1-[1,1-dimethyl-3-(4-sulphonatobutyl)benz[e]indolium-2-yl]-1,3-pentadiene sodium salt, 2-[5-[1,1-Dimethyl-3-(4-sulphobutyl)-1,3-dihydrobenzo[e]indol-2-ylidene]-1,3-pentadienyl]-1,1-dimethyl-3-(4-sulphobutyl)-1H-benzo[e]indolium hydroxide Inner salt sodium salt

D4463-1G D4463-5G
TCIAD4463-1GEA 102 EUR
TCIAD4463-1G TCIAD4463-5G
3,3,3',3'-Tetramethyl-1,1'-bis(4-sulphobutyl)benzoindodicarbocyanine sodium salt ≥98.0% (by HPLC, total nitrogen)
3,3,3',3'-Tetramethyl-1,1'-bis(4-sulphobutyl)benzoindodicarbocyanine sodium salt
Formula: C₄₁H₄₅N₂NaO₆S₂
MW: 748,93 g/mol
Número MDL: MFCD16621036
Núm. CAS: 64285-36-5

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Absorbance (E1%1cm) min. 2800 (MeOH, 675 to 685nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR