1,3-Dibromo-2,2-dimetoxipropano ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
D4579-5G
D4579-25G
TCIAD4579-5GEA
43
EUR
TCIAD4579-5G
TCIAD4579-25G
1,3-Dibromo-2,2-dimetoxipropano ≥98.0% (por GC)
1,3-Dibromo-2,2-dimetoxipropano
Formula:
BrH₂CC(OCH₃)₂CH₂Br MW: 261,94 g/mol |
Número MDL:
MFCD00051792 Núm. CAS: 22094-18-4 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 64.0 to 68.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...