3,5-Dimethylisoxazole-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3,5-Dimethylisoxasole-4-carboxylic acid
D4603-1G
D4603-5G
TCIAD4603-1GEA
36
EUR
TCIAD4603-1G
TCIAD4603-5G
3,5-Dimethylisoxazole-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
3,5-Dimethylisoxazole-4-carboxylic acid
Formula:
C₆H₇NO₃ MW: 141,13 g/mol |
Número MDL:
MFCD00051657 Núm. CAS: 2510-36-3 EINECS: 219-724-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...