Dimethyl-bis-(phenylethynyl)silane ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
Dimethylbis(phenylethynyl)silane
D4625-1G
D4625-5G
TCIAD4625-1GEA
41
EUR
TCIAD4625-1G
TCIAD4625-5G
Dimethyl-bis-(phenylethynyl)silane ≥98.0% (por GC)
Dimethyl-bis-(phenylethynyl)silane
Formula:
C₁₈H₁₆Si MW: 260,41 g/mol Pto. Ebullición: 177°C |
Número MDL:
MFCD00270967 Núm. CAS: 2170-08-3 |
Resultados test especificaciones
Purity (GC) | min. 98.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...