Diethylene glycol butyl methyl ether ≥99.0% (por GC)
Proveedor: TCI
Sinónimos:
1-[2-(2-Methoxyethoxy)ethoxy]butane
D4676-25ML
D4676-500ML
TCIAD4676-25MLEA
16
EUR
TCIAD4676-25ML
TCIAD4676-500ML
Diethylene glycol butyl methyl ether ≥99.0% (por GC)
Diethylene glycol butyl methyl ether
Formula:
C₉H₂₀O₃ MW: 176,26 g/mol |
Número MDL:
MFCD00518661 Núm. CAS: 7382-32-3 EINECS: 230-954-5 |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...