4-(Diethylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 4-(Diethylcarbamoyl)benzeneboronic acid, 4-(N,N-Diethylaminocarbonyl)phenylboronic acid

D4681-1G D4681-5G
TCIAD4681-1GEA 83 EUR
TCIAD4681-1G TCIAD4681-5G
4-(Diethylcarbamoyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Diethylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
MW: 221,06 g/mol
Número MDL: MFCD03411949
Núm. CAS: 389621-80-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR