2,6-Dimethoxy-3-pyridineboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid
D4690-1G
D4690-5G
TCIAD4690-1GEA
35
EUR
TCIAD4690-1G
TCIAD4690-5G
2,6-Dimethoxy-3-pyridineboronic acid (contains varying amounts of Anhydride)
2,6-Dimethoxy-3-pyridineboronic acid
Formula:
C₇H₁₀BNO₄ MW: 182,97 g/mol Pto. de fusión: 109°C |
Número MDL:
MFCD03788239 Núm. CAS: 221006-70-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...