3,5-Dihydroxy-4-methylbenzoic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

D4712-1G D4712-5G
TCIAD4712-1GEA 31 EUR
TCIAD4712-1G TCIAD4712-5G
3,5-Dihydroxy-4-methylbenzoic acid ≥98.0% (by titrimetric analysis)
3,5-Dihydroxy-4-methylbenzoic acid
Formula: C₈H₈O₄
MW: 168,15 g/mol
Número MDL: MFCD00002514
Núm. CAS: 28026-96-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR