3,5-Dibromo-p-toluidine ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3,5-Dibromo-4-methylaniline
D4730-1G
D4730-5G
TCIAD4730-1GEA
111
EUR
TCIAD4730-1G
TCIAD4730-5G
3,5-Dibromo-p-toluidine ≥97.0% (by GC, titration analysis)
3,5-Dibromo-p-toluidine
Formula:
C₇H₇Br₂N MW: 264,96 g/mol |
Número MDL:
MFCD00151806 Núm. CAS: 13194-73-5 |
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...