Diphenhydramine ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(Benzhydryloxy)-N,N-dimethylethylamine, 2-(Benzhydryloxy)-N,N-dimethylethanamine, 2-Diphenylmethoxy-N,N-dimethylethylamine

D4744-5G D4744-25G
TCIAD4744-5GEA 25 EUR
TCIAD4744-5G TCIAD4744-25G
Diphenhydramine ≥98.0% (by GC, titration analysis)
Diphenhydramine
Formula: C₁₇H₂₁NO
MW: 255,36 g/mol
Número MDL: MFCD00274173
Núm. CAS: 58-73-1
EINECS: 200-396-7
Índice Merck: 14,03309

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR