4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole ≥98.0% (por GC)

Proveedor: TCI

D4750-200MG D4750-1G
TCIAD4750-200MGEA 40 EUR
TCIAD4750-200MG TCIAD4750-1G
4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole ≥98.0% (por GC)
4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole
Formula: C₆HBr₂FN₂S
MW: 311,95 g/mol
Número MDL: MFCD26939241
Núm. CAS: 1347736-74-6

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR