2-Difluoromethyl-1H-benzoimidazole ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(Difluoromethyl)-1H-benzo[d]imidazole, 2-(Difluoromethyl)benzimidazole
D4763-1G
D4763-5G
TCIAD4763-1GEA
79
EUR
TCIAD4763-1G
TCIAD4763-5G
2-Difluoromethyl-1H-benzoimidazole ≥98.0% (by GC, titration analysis)
2-Difluoromethyl-1H-benzoimidazole
Formula:
C₈H₆F₂N₂ MW: 168,15 g/mol |
Número MDL:
MFCD00463015 Núm. CAS: 705-09-9 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...