N,N'-Di(1-naphthyl)benzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Proveedor: TCI
Sinónimos:
N,N’-Di(1-naphthyl)-4,4’-benzidine
D4768-1G
D4768-5G
TCIAD4768-1GEA
49
EUR
TCIAD4768-1G
TCIAD4768-5G
N,N'-Di(1-naphthyl)benzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
N,N'-Di(1-naphthyl)benzidine
Formula:
C₃₂H₂₄N₂ |
Número MDL:
MFCD09261380 Núm. CAS: 152670-41-2 |
Resultados test especificaciones
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...