4-(Difluoromethoxy)benzenesulphonamide ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

D4778-1G D4778-5G
TCIAD4778-1GEA 51 EUR
TCIAD4778-1G TCIAD4778-5G
4-(Difluoromethoxy)benzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
4-(Difluoromethoxy)benzenesulphonamide
Formula: C₇H₇F₂NO₃S
MW: 223,2000 g/mol
Número MDL: MFCD03407973
Núm. CAS: 874781-09-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 110.0 to 115.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR