N,N'-Diphenyl-N,N'-di(p-tolyl)benzidine ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
N,N'-Bis(4-methylphenyl)-N,N'-diphenylbenzidine, 4,4'-Bis[N-phenyl-N-(p-tolyl)amino]biphenyl
D4834-200MG
D4834-1G
TCIAD4834-200MGEA
81
EUR
TCIAD4834-200MG
TCIAD4834-1G
N,N'-Diphenyl-N,N'-di(p-tolyl)benzidine ≥98.0% (por GC)
N,N'-Diphenyl-N,N'-di(p-tolyl)benzidine
Formula:
C₃₈H₃₂N₂ MW: 516.69 g/mol |
Número MDL:
MFCD00799301 Núm. CAS: 20441-06-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...