1-(2,4-Difluorofenil)-2-tiourea ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Peligro
|
Sinónimos:
N-(2,4-Difluorofenil)tiourea, 2,4-Difluorofeniltiourea
D4861-1G
D4861-5G
TCIAD4861-1GEA
18
EUR
TCIAD4861-1G
TCIAD4861-5G
1-(2,4-Difluorofenil)-2-tiourea ≥98.0% (by HPLC, total nitrogen)
1-(2,4-Difluorofenil)-2-tiourea
Formula:
(C₆H₃F₂)NC(S)NH₂ MW: 188,2 g/mol |
Número MDL:
MFCD00041150 Núm. CAS: 175277-76-6 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...