2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene ≥99.0% (by HPLC)
Proveedor: TCI
D4862-200MG
D4862-1G
TCIAD4862-200MGEA
130
EUR
TCIAD4862-200MG
TCIAD4862-1G
2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene ≥99.0% (by HPLC)
2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene
Formula:
C₃₄H₃₂Br₂ |
Núm. CAS:
1187763-68-3 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...