3,4-Difluorobencenosulfonamida ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
D4892-1G
D4892-5G
TCIAD4892-1GEA
17
EUR
TCIAD4892-1G
TCIAD4892-5G
3,4-Difluorobencenosulfonamida ≥98.0% (por GC)
3,4-Difluorobencenosulfonamida
Formula:
C₆H₅F₂NO₂S MW: 193,17 g/mol |
Número MDL:
MFCD00728800 Núm. CAS: 108966-71-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 95.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...