2,4-Dichloronicotinc acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
D4901-1G
D4901-5G
TCIAD4901-1GEA
35
EUR
TCIAD4901-1G
TCIAD4901-5G
2,4-Dichloronicotinc acid ≥98.0% (by GC, titration analysis)
2,4-Dichloronicotinc acid
Formula:
C₆H₃Cl₂NO₂ |
Número MDL:
MFCD06796292 Núm. CAS: 262423-77-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...