3,3''-Di(9H-carbazol-9-yl)-1,1':3',1''-terphenyl ≥97.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: 3,3''-Di(9H-carbazol-9-yl)-m-terphenyl

D4904-200MG D4904-1G
TCIAD4904-200MGEA 109 EUR
TCIAD4904-200MG TCIAD4904-1G
3,3''-Di(9H-carbazol-9-yl)-1,1':3',1''-terphenyl ≥97.0% (by HPLC, total nitrogen)
3,3''-Di(9H-carbazol-9-yl)-1,1':3',1''-terphenyl
Formula: C₄₂H₂₈N₂
MW: 560,70 g/mol
Núm. CAS: 1116499-73-0

Order Now

Resultados test especificaciones

Appearance White to Light gray to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR