9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
D4919-200MG
D4919-1G
TCIAD4919-200MGEA
72
EUR
TCIAD4919-200MG
TCIAD4919-1G
9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
9-[3-(Dibenzofuran-2-yl)phenyl]-9H-carbazole
Formula:
C₃₀H₁₉NO |
Número MDL:
MFCD30478530 Núm. CAS: 1338446-77-7 |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...