1-(2,6-Diisopropylphenyl)imidazole ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
D4948-1G
D4948-5G
TCIAD4948-1GEA
119
EUR
TCIAD4948-1G
TCIAD4948-5G
1-(2,6-Diisopropylphenyl)imidazole ≥98.0% (by titrimetric analysis)
1-(2,6-Diisopropylphenyl)imidazole
Formula:
C₁₅H₂₀N₂ Pto. Ebullición: 105°C |
Número MDL:
MFCD13299721 Núm. CAS: 25364-47-0 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...