6,6'-Dibromo-9,9'-diphenyl-3,3'-bicarbazole ≥95.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

D4980-200MG D4980-1G
TCIAD4980-200MGEA 91 EUR
TCIAD4980-200MG TCIAD4980-1G
6,6'-Dibromo-9,9'-diphenyl-3,3'-bicarbazole ≥95.0% (by HPLC, total nitrogen)
6,6'-Dibromo-9,9'-diphenyl-3,3'-bicarbazole
Formula: C₃₆H₂₂Br₂N₂
MW: 642,39 g/mol
Número MDL: MFCD30749258
Núm. CAS: 354135-75-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96f75a79a4819a2bbe22de87c87c0ff9937596f0&vid=96f75a79a4819a2bbe22de87c87c0ff9937596f0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR