2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 9,9-Diphenyl-9H-fluorene-4-boronic acid Pinacol ester, 9,9-Diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene

D5033-200MG D5033-1G
TCIAD5033-200MGEA 49 EUR
TCIAD5033-200MG TCIAD5033-1G
2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₃₁H₂₉BO₂
MW: 444,38 g/mol
Número MDL: MFCD30187268
Núm. CAS: 1259280-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07dfc055f0736ce6d797e36f61f1747ddc002415&vid=07dfc055f0736ce6d797e36f61f1747ddc002415&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR