2,5-Dichloro-N-[2-(isopropylsulphonyl)phenyl]pyrimidin-4-amine ≥97.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
2,5-Dichloro-N-(2-(isopropylsulphonyl)phenyl)pyrimidin-4-amine
D5068-200MG
D5068-1G
TCIAD5068-200MGEA
42
EUR
TCIAD5068-200MG
TCIAD5068-1G
2,5-Dichloro-N-[2-(isopropylsulphonyl)phenyl]pyrimidin-4-amine ≥97.0% (by HPLC, total nitrogen)
2,5-Dichloro-N-[2-(isopropylsulphonyl)phenyl]pyrimidin-4-amine
Formula:
C₁₃H₁₃Cl₂N₃O₂S |
Número MDL:
MFCD12827956 Núm. CAS: 761440-16-8 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 149.0 to 153.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...