2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 1,7'-Dimethyl-2'-propyl-2,5'-bibenzimidazole

D5079-25G D5079-5G
TCIAD5079-25GEA 226 EUR
TCIAD5079-25G TCIAD5079-5G
2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole ≥98.0% (by HPLC)
2-N-Propyl-4-methyl-6-(1'-methylbenzimidazole-2-yl)benzimidazole
Formula: C₁₉H₂₀N₄
MW: 304,397 g/mol
Número MDL: MFCD09029193
Núm. CAS: 152628-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f589beb000be8887b3a7fed388870064b140c53c&vid=c942fb2776152db2317a176b7167f5e33cc183d7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR