3,5-Dimethyl-4-propoxyphenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
(3,5-Dimethyl-4-propoxyphenyl)boronic acid
D5080-1G
D5080-5G
TCIAD5080-1GEA
20
EUR
TCIAD5080-1G
TCIAD5080-5G
3,5-Dimethyl-4-propoxyphenylboronic acid (contains varying amounts of Anhydride)
3,5-Dimethyl-4-propoxyphenylboronic acid
Formula:
C₁₁H₁₇BO₃ |
Número MDL:
MFCD05865178 Núm. CAS: 357611-51-9 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...