2,3-Difluoro-4-n-propoxybenzeneboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: (2,3-Difluoro-4-propoxyphenyl)boronic acid, 2,3-Difluoro-4-propoxyphenylboronic acid, 2,3-Difluoro-4-propoxybenzeneboronic acid

D5083-1G D5083-5G
TCIAD5083-1GEA 50 EUR
TCIAD5083-1G TCIAD5083-5G
2,3-Difluoro-4-n-propoxybenzeneboronic acid (contains varying amounts of Anhydride)
2,3-Difluoro-4-n-propoxybenzeneboronic acid
Formula: C₉H₁₁BF₂O₃
MW: 215.991 g/mol
Número MDL: MFCD14584457
Núm. CAS: 212837-49-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24e48a3d4605068112fd408ad21573836a5c618e&vid=24e48a3d4605068112fd408ad21573836a5c618e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR