4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl ≥98.0% (por GC)
Proveedor: TCI
D5129-1G
D5129-5G
TCIAD5129-1GEA
39
EUR
TCIAD5129-1G
TCIAD5129-5G
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl ≥98.0% (por GC)
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl
Formula:
C₂₁H₁₃F₇O |
Número MDL:
MFCD22376741 Núm. CAS: 303186-19-8 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 67.0 to 72.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...