2,6-Dichloro-3-nitrobenzoic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

D5173-1G D5173-5G
TCIAD5173-1GEA 68 EUR
TCIAD5173-1G TCIAD5173-5G
2,6-Dichloro-3-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
2,6-Dichloro-3-nitrobenzoic acid
Formula: C₇H₃Cl₂NO₄
MW: 236,0100 g/mol
Número MDL: MFCD00075422
Núm. CAS: 55775-97-8

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR