4,5-Dibromo-1H-1,2,3-triazole ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
D5205-1G
D5205-5G
TCIAD5205-1GEA
43
EUR
TCIAD5205-1G
TCIAD5205-5G
4,5-Dibromo-1H-1,2,3-triazole ≥98.0% (by titrimetric analysis)
4,5-Dibromo-1H-1,2,3-triazole
Formula:
C₂HBr₂N₃ |
Número MDL:
MFCD08062401 Núm. CAS: 15294-81-2 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...