2,5-Dibromo-3-hexadecylthiophene ≥96.0% (por GC)
Proveedor: TCI
D5242-1G
D5242-5G
TCIAD5242-1GEA
79
EUR
TCIAD5242-1G
TCIAD5242-5G
2,5-Dibromo-3-hexadecylthiophene ≥96.0% (por GC)
2,5-Dibromo-3-hexadecylthiophene
Formula:
C₂₀H₃₄Br₂S MW: 466,36 g/mol Temperatura de almacenaje: Nevera |
Núm. CAS:
178452-13-6 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 34.0 to 38.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...